Skip to Content
MilliporeSigma
Quercetin 3-O-rutinoside-7-O-glucoside phyproof® Reference Substance

Quercetin 3-O-rutinoside-7-O-glucoside

Quercetin 3-O-rutinoside-7-O-glucoside phyproof® Reference Substance
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside 7-glucoside, 3-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-7-(β-D-glucopyranosyloxy)-5-hydroxy-4H-1-benzopyran-4-one, Quercetin 3-rhamnoglucoside 7-glucoside
Empirical Formula (Hill Notation):
C33H40O21
CAS Number:
Molecular Weight:
772.66

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing