Skip to Content
MilliporeSigma
Pyriproxyfen PESTANAL®, analytical standard

Pyriproxyfen

Pyriproxyfen PESTANAL®, analytical standard
Synonym(s):
2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine
Empirical Formula (Hill Notation):
C20H19NO3
CAS Number:
Molecular Weight:
321.37
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing