Skip to Content
MilliporeSigma
Pyridinium p-toluenesulfonate puriss., ≥99.0% (T)

Pyridinium p-toluenesulfonate

Pyridinium p-toluenesulfonate puriss., ≥99.0% (T)
Synonym(s):
p-Toluenesulfonic acid pyridine salt, PPTS, Pyridine p-toluenesulfonate
Empirical Formula (Hill Notation):
C12H13NO3S
CAS Number:
Molecular Weight:
251.30
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing