Skip to Content
MilliporeSigma
Proton-sponge® 99%

Proton-sponge®

Proton-sponge® 99%
Synonym(s):
1,8-Bis(dimethylamino)naphthalene, N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine
Linear Formula:
C10H6[N(CH3)2]2
CAS Number:
Molecular Weight:
214.31
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing