Skip to Content
MilliporeSigma
Propionamide 97%

Propionamide

Propionamide 97%
Synonym(s):
Propanamide, Propylamide, Propionamide
Linear Formula:
CH3CH2CONH2
CAS Number:
Molecular Weight:
73.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing