Skip to Content
MilliporeSigma
Proguanil Related Compound C pharmaceutical secondary standard, certified reference material

Proguanil Related Compound C

Proguanil Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
1,5-Bis(4-chlorophenyl)biguanide monohydrochloride monohydrochloride, N,N′-Bis(4-chlorophenyl)imidodicarbonimidic diamide monohydrochloride
Empirical Formula (Hill Notation):
C14H13Cl2N5 · HCl
CAS Number:
Molecular Weight:
358.65

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade