Skip to Content
MilliporeSigma
Prenyl benzoate ≥98%, FG

Prenyl benzoate

Prenyl benzoate ≥98%, FG
Synonym(s):
3-Methyl-2-butenyl benzoate
Empirical Formula (Hill Notation):
C12H14O2
CAS Number:
Molecular Weight:
190.24

Products

Product Category
Brand
Formula Weight
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing