Skip to Content
MilliporeSigma
vibrant-m

Pravastatin impurity A

vibrant-m
Synonym(s):
(3R,5R)-3,5-Dihydroxy-7-[(1S,2S,6R,8S,8aR)-6-hydroxy-2-methyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]heptanoic acid
Empirical Formula (Hill Notation):
C23H36O7
CAS Number:
Molecular Weight:
424.53

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade