Skip to Content
MilliporeSigma
Potassium Cyanide Pharmaceutical Secondary Standard; Certified Reference Material

Potassium cyanide

Potassium Cyanide Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Potassium cyanide
Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Package Size
Special Grade