Skip to Content
MilliporeSigma
Potassium bis(trimethylsilyl)amide 95%

Potassium bis(trimethylsilyl)amide

Potassium bis(trimethylsilyl)amide 95%
Synonym(s):
Hexamethyldisilazane potassium salt, Potassium hexamethyldisilazide
Linear Formula:
[(CH3)3Si]2NK
CAS Number:
Molecular Weight:
199.48

Products

Product Category
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing