Skip to Content
MilliporeSigma
Potassium aminetrichloroplatinate(II) 99.9% trace metals basis

Potassium aminetrichloroplatinate(II)

Potassium aminetrichloroplatinate(II) 99.9% trace metals basis
Synonym(s):
Magnus red anion salt, Potassium trichloroammineplatinate(II)
Linear Formula:
KPt(NH3)Cl3
CAS Number:
Molecular Weight:
357.57

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing