Skip to Content
MilliporeSigma
Poly(styrene)-block-poly(acrylic acid) PS:PAA 30,000:2,000, PDI ≤1.1

Polystyrene-block-poly(acrylic acid)

Poly(styrene)-block-poly(acrylic acid) PS:PAA 30,000:2,000, PDI ≤1.1
Synonym(s):
Poly(styrene)-block-poly(acrylic acid), PS-PAA, PS-PAA amphiphilic diblock copolymer
Linear Formula:
C4H6N(C8H8)x(C3H4O2)yH

Products

Brand
Melting Point (°C)
Physical Form
Polymer Composition
Polymer Type
Showing 1-2 of 2