Skip to Content
MilliporeSigma
Polysorbate 20 certified reference material, pharmaceutical secondary standard

Polysorbate 20

Polysorbate 20 certified reference material, pharmaceutical secondary standard
Synonym(s):
TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20
CAS Number:

Products

Product Category
Brand
Biological Source
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade