Skip to Content
MilliporeSigma
Poly(sodium 4-styrenesulfonate) average Mw ~1,000,000, powder

Poly(sodium 4-styrenesulfonate)

Poly(sodium 4-styrenesulfonate) average Mw ~1,000,000, powder
Synonym(s):
PSS, Poly(4-styrenesulfonic acid) sodium salt
Linear Formula:
(C8H7NaO3S)n
CAS Number:

Products

Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-2 of 2