Skip to Content
MilliporeSigma
Polypropylene-graft-maleic anhydride viscosity 425 cPs

Polypropylene-graft-maleic anhydride

Polypropylene-graft-maleic anhydride viscosity 425 cPs
Synonym(s):
maleated copolymer, maleated polymer, maleic copolymer
CAS Number:

Products

Melting Point (°C)
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing