Skip to Content
MilliporeSigma
Polyoxyethylene bis(azide) average Mn 2,000

Polyoxyethylene bis(azide)

Polyoxyethylene bis(azide) average Mn 2,000
Synonym(s):
O,O′-Bis(2-azidoethyl)polyethylene glycol, PEG-diazide
Linear Formula:
N3CH2CH2(OCH2CH2)nN3
CAS Number:

Products

Brand
Melting Point (°C)
Quality Segment
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing