Skip to Content
MilliporeSigma
Poly(methylhydrosiloxane) viscosity 15-40 mPa.s (20 °C)

Poly(methylhydrosiloxane)

Poly(methylhydrosiloxane) viscosity 15-40 mPa.s (20 °C)
Synonym(s):
Methylhydrogensiloxane polymer, PMHS
Linear Formula:
(CH3)3SiO[(CH3)HSiO]nSi(CH3)3
CAS Number:

Products

Sustainability
Feature
Physical Form
Quality Segment
Showing 1-2 of 2