Skip to Content
MilliporeSigma
Poly(isobutylene-alt-maleic anhydride) average Mw ~6,000, 12-200 mesh (85%)

Poly(isobutylene-alt-maleic anhydride)

Poly(isobutylene-alt-maleic anhydride) average Mw ~6,000, 12-200 mesh (85%)
Synonym(s):
maleated copolymer, maleated polymer, maleic copolymer
Linear Formula:
(C4H2O3 · C4H8)X
CAS Number:

Products

Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing