Skip to Content
MilliporeSigma
Poly(ethylene glycol) diamine average Mn 6,000

Poly(ethylene glycol) diamine

Poly(ethylene glycol) diamine average Mn 6,000
Synonym(s):
Poly(ethylene glycol) bis(amine), O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)
Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:

Products

Brand
Color
Melting Point (°C)
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type