Skip to Content
MilliporeSigma
Poly(D,L-lactide-co-glycolide) lactide:glycolide (50:50), ester terminated, average Mw 100,000

Poly(D,L-lactide-co-glycolide)

Poly(D,L-lactide-co-glycolide) lactide:glycolide (50:50), ester terminated, average Mw 100,000
Synonym(s):
PLGA
Linear Formula:
[C3H4O2]x[C2H2O2]y

Products

Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Optical Isomer
Polymer Composition
Polymer Type