Skip to Content
MilliporeSigma
Polycaprolactone average Mn 45,000

Polycaprolactone

Polycaprolactone average Mn 45,000
Synonym(s):
2-Oxepanone homopolymer, 6-Caprolactone polymer
Linear Formula:
(C6H10O2)n

Products

Brand
Melting Point (°C)
Physical Form
Quality Segment
Application
Polymer Composition
Polymer Type
Showing 1-3 of 3