Skip to Content
MilliporeSigma
Phosphazene base P2-Et ≥98.0% (NT)

Phosphazene base P2-Et

Phosphazene base P2-Et ≥98.0% (NT)
Synonym(s):
1-Ethyl-2,2,4,4,4-pentakis(dimethylamino)-2λ5,4λ5-catenadi(phosphazene), Tetramethyl(tris(dimethylamino)phosphoranylidene)phosphorictriamid-Et-imin
Linear Formula:
[(CH3)2N]3P=NP(=NC2H5)[N(CH3)2]2
CAS Number:
Molecular Weight:
339.40

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing