Skip to Content
MilliporeSigma
Phenol BioUltra, ≥99.5% (GC)

Phenol

Phenol BioUltra, ≥99.5% (GC)
Synonym(s):
Hydroxybenzene
Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique