Skip to Content
MilliporeSigma
Pentafluorobenzene 98%

Pentafluorobenzene

Pentafluorobenzene 98%
Linear Formula:
HC6F5
CAS Number:
Molecular Weight:
168.06
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing