Skip to Content
MilliporeSigma
p-Coumaric acid primary reference standard

p-Coumaric acid

p-Coumaric acid primary reference standard
Synonym(s):
trans-4-Hydroxycinnamic acid
Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing