Skip to Content
MilliporeSigma
p-Bromo-DL-phenylalanine 99%

p-Bromo-DL-phenylalanine

p-Bromo-DL-phenylalanine 99%
Linear Formula:
BrC6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
244.09
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing