Skip to Content
MilliporeSigma
PBFI-AM suitable for fluorescence, ≥98.0% (HPCE)

PBFI-AM

PBFI-AM suitable for fluorescence, ≥98.0% (HPCE)
Synonym(s):
Potassium indicator PBFI-AM, Potassium-binding benzofuran isophthalate-AM
Empirical Formula (Hill Notation):
C58H62N2O24
CAS Number:
Molecular Weight:
1171.11

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing