Skip to Content
MilliporeSigma
Papaverine hydrochloride British Pharmacopoeia (BP) Reference Standard

Papaverine hydrochloride

Papaverine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride
Empirical Formula (Hill Notation):
C20H21NO4 · HCl
CAS Number:
Molecular Weight:
375.85
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique