Skip to Content
MilliporeSigma
Pamoic acid United States Pharmacopeia (USP) Reference Standard

Pamoic acid

Pamoic acid United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,1′-Methylene-bis(2-hydroxy-3-naphthoic acid), 4,4′-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid
Empirical Formula (Hill Notation):
C23H16O6
CAS Number:
Molecular Weight:
388.37

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing