
Paclitaxel Related Compound B

Synonym(s):
10-Deacetyl-7-epipaclitaxel, 7-epi-10-Deacetyltaxol
Empirical Formula (Hill Notation):
C45H49NO13
CAS Number:
Molecular Weight:
811.87
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2