Skip to Content
MilliporeSigma
Osmium tetroxide Sealed ampule.

Osmium tetroxide

Osmium tetroxide Sealed ampule.
Synonym(s):
Osmium(VIII)-oxide, ‘Osmic acid’
Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Reagent Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing