Skip to Content
MilliporeSigma
Olopatadine Hydrochloride pharmaceutical secondary standard, certified reference material

Olopatadine hydrochloride

Olopatadine Hydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
(11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid hydrochloride, AL-4943A, KW-4679, Opatanol, Patanol, Olopatadine hydrochloride
Empirical Formula (Hill Notation):
C21H23NO3·HCl
CAS Number:
Molecular Weight:
373.87

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade