Skip to Content
MilliporeSigma
Norepinephrine Bitartrate certified reference material, pharmaceutical secondary standard

Norepinephrine bitartrate

Norepinephrine Bitartrate certified reference material, pharmaceutical secondary standard
Synonym(s):
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, L-(−)-Noradrenaline (+)-bitartrate salt monohydrate
Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade