Skip to Content
MilliporeSigma
Nonafluorobutyl methyl ether puriss., ≥99.0% (total assay of isomers, GC)

Nonafluorobutyl methyl ether

Nonafluorobutyl methyl ether puriss., ≥99.0% (total assay of isomers, GC)
Synonym(s):
HFE-7100, Methoxynonafluorobutane, Methoxyperfluorobutane, Nonafluoromethoxybutane
Linear Formula:
CH3OCF2R, R = -CF2CF2CF3 or -CF(CF3)2
CAS Number:
Molecular Weight:
250.06

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing