Skip to Content
MilliporeSigma
N,N-Dimethylethylamine ≥99%

N,N-Dimethylethylamine

N,N-Dimethylethylamine ≥99%
Synonym(s):
N-Ethyldimethylamine, DMEA, N,N-Dimethylethylamine
Linear Formula:
(CH3)2NC2H5
CAS Number:
Molecular Weight:
73.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing