
Nitrogen and oxygen isotopes in nitrate

Synonym(s):
Sodium nitrate, Chile salpeter
Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing