
Nitisinone

Synonym(s):
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 2-[2-Nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione, NTBC, Nitisone, SC 0735, Nitisinone
Empirical Formula (Hill Notation):
C14H10F3NO5
CAS Number:
Molecular Weight:
329.23
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing