Skip to Content
MilliporeSigma
Nitazoxanide Related Compound A pharmaceutical secondary standard, certified reference material

Nitazoxanide Related Compound A

Nitazoxanide Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
2-Amino-5-nitrothiazole
Empirical Formula (Hill Notation):
C3H3N3O2S
CAS Number:
Molecular Weight:
145.14

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade