
Nimodipine impurity A

Synonym(s):
2-Methoxyethyl 1-methylethyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate
Empirical Formula (Hill Notation):
C21H24N2O7
CAS Number:
Molecular Weight:
416.42
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing