Skip to Content
MilliporeSigma
Nicotinamide British Pharmacopoeia (BP) Reference Standard

Nicotinamide

Nicotinamide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP, Nicotinamide
Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique