Skip to Content
MilliporeSigma
N-Hexylmethylamine 96%

N-Hexylmethylamine

N-Hexylmethylamine 96%
Synonym(s):
N-Methylhexylamine
Linear Formula:
CH3NH(CH2)5CH3
CAS Number:
Molecular Weight:
115.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing