Skip to Content
MilliporeSigma
NG-Methyl-L-arginine acetate salt ≥98% (TLC)

NG-Methyl-L-arginine acetate salt

NG-Methyl-L-arginine acetate salt ≥98% (TLC)
Synonym(s):
Nω-Methyl-L-arginine acetate salt, L-NMMA, N5-(Methylamidino)-L-ornithine acetate salt, NG-Monomethyl-L-arginine acetate salt
Empirical Formula (Hill Notation):
C7H16N4O2 · C2H4O2
CAS Number:
Molecular Weight:
248.28

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1