Skip to Content
MilliporeSigma
Neohesperidin pharmaceutical secondary standard, certified reference material

Neohesperidin

Neohesperidin pharmaceutical secondary standard, certified reference material
Synonym(s):
(S)-4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Hesperetin 7-O-neohesperidoside, NSC 31048, Neohesperidin
Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade