Skip to Content
MilliporeSigma
Neodymium foil, thickness 0.1 mm, size 50 × 50 mm, purity 99%

Neodymium

Neodymium foil, thickness 0.1 mm, size 50 × 50 mm, purity 99%
Synonym(s):
Neodymium
Empirical Formula (Hill Notation):
Nd
CAS Number:
Molecular Weight:
144.24

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Showing 1-17 of 17
Compare
Product Number
Description
Pricing