Skip to Content
MilliporeSigma
Nefopam hydrochloride ≥98% (HPLC)

Nefopam hydrochloride

Nefopam hydrochloride ≥98% (HPLC)
Synonym(s):
3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine hydrochloride, 5-Methyl-1-phenyl-1,3,4,6-tetrahydro-5H-benz[f]-2,5-oxazocine hydrochloride
Empirical Formula (Hill Notation):
C17H19NO · HCl
CAS Number:
Molecular Weight:
289.80
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing