Skip to Content
MilliporeSigma
N-Boc-ethanolamine 98%

N-Boc-ethanolamine

N-Boc-ethanolamine 98%
Synonym(s):
N-(tert-Butoxycarbonyl)ethanolamine, tert-Butyl N-(2-hydroxyethyl)carbamate, Boc-2-aminoethanol, Boc-glycinol
Linear Formula:
(CH3)3COCONHCH2CH2OH
CAS Number:
Molecular Weight:
161.20

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing