Skip to Content
MilliporeSigma
N-Benzyl-L-prolinol 99%

N-Benzyl-L-prolinol

N-Benzyl-L-prolinol 99%
Synonym(s):
(S)-(−)-1-Benzylpyrrolidine-2-methanol
Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing