Skip to Content
MilliporeSigma
N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate ≥98% (TLC)

N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate

N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate ≥98% (TLC)
Synonym(s):
Adjuvant Peptide, Muramyl dipeptide
Empirical Formula (Hill Notation):
C19H32N4O11 · xH2O
CAS Number:
Molecular Weight:
492.48 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1