Skip to Content
MilliporeSigma
N-(4-Aminobutyl)-N-ethylisoluminol ≥90%

N-(4-Aminobutyl)-N-ethylisoluminol

N-(4-Aminobutyl)-N-ethylisoluminol ≥90%
Synonym(s):
4-(N1-Ethyl-4-aminobutylamino)phthalic hydrazide, 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI
Empirical Formula (Hill Notation):
C14H20N4O2
CAS Number:
Molecular Weight:
276.33

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing