Skip to Content
MilliporeSigma
Myrcene analytical standard

Myrcene

Myrcene analytical standard
Synonym(s):
β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene
Linear Formula:
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing